2-Chloro-5-methoxy-4-pyrimidinamine 97%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-5-methoxypyrimidin-4-amine

OR926836-5G
APOSOR926836-5GEA 62.5 EUR
APOSOR926836-5G
2-Chloro-5-methoxy-4-pyrimidinamine 97%
2-Chloro-5-methoxy-4-pyrimidinamine
Formel: C₅H₆ClN₃O
Molecular Weight: 159,574 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17169913
CAS-Nummer: 99979-77-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR