6-Methylchromon Hydrat 95%

Lieferant: Apollo Scientific

OR927025-1G OR927025-5G
APOSOR927025-1GEA 132 EUR
APOSOR927025-1G APOSOR927025-5G
6-Methylchromon Hydrat 95%
6-Methylchromon Hydrat
Formel: C₁₀H₈O₂·xH₂O
Molecular Weight: 160,17 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00209598
CAS-Nummer: 314041-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR