6-Chloro-4-formyl-nicotinic acid 95%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-4-formylnicotinic acid

OR927158-1G OR927158-250MG
APOSOR927158-1GEA 857 EUR
APOSOR927158-1G APOSOR927158-250MG
6-Chloro-4-formyl-nicotinic acid 95%
6-Chloro-4-formyl-nicotinic acid
Formel: C₇H₄ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18258739
CAS-Nummer: 1031433-06-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR