Propyl-(tetrahydro-pyran-4-yl)-amine 98%
Lieferant: Apollo Scientific
OR927201-1G
OR927201-250MG
APOSOR927201-1GEA
435
EUR
APOSOR927201-1G
APOSOR927201-250MG
Propyl-(tetrahydro-pyran-4-yl)-amine 98%
Propyl-(tetrahydro-pyran-4-yl)-amine
Formel:
C₈H₁₇NO Molecular Weight: 143,227 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06658506 CAS-Nummer: 192811-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...