1-(4-Hydroxy-7-methyl-indan-5-yl)-ethanone 95%

Lieferant: Apollo Scientific

OR927212-1G OR927212-250MG
APOSOR927212-1GEA 754 EUR
APOSOR927212-1G APOSOR927212-250MG
1-(4-Hydroxy-7-methyl-indan-5-yl)-ethanone 95%
1-(4-Hydroxy-7-methyl-indan-5-yl)-ethanone
Formel: C₁₂H₁₄O₂
Molecular Weight: 190.238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173684
CAS-Nummer: 175136-13-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR