4-Propylbenzylamine 95%
Lieferant: Apollo Scientific
Synonyme:
(4-Propylphenyl)methanamine
OR927272-1G
OR927272-250MG
APOSOR927272-1GEA
162
EUR
APOSOR927272-1G
APOSOR927272-250MG
4-Propylbenzylamine 95%
4-Propylbenzylamine
Formel:
C₁₀H₁₅N Molecular Weight: 149,233 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411009 CAS-Nummer: 538342-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...