Methyl 3-(ethylamino)propanoate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl N-ethyl-β-alaninate, 3-Ethylamino-propionic acid methyl ester

OR927277-1G OR927277-5G
APOSOR927277-1GEA 256 EUR
APOSOR927277-1G APOSOR927277-5G
Methyl 3-(ethylamino)propanoate 97%
Methyl 3-(ethylamino)propanoate
Formel: C₆H₁₃NO₂
Molecular Weight: 131,173 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09035843
CAS-Nummer: 3440-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR