Ethyl 3-(4-pyridyl)propiolate 95%

Lieferant: Apollo Scientific

OR927308-1G OR927308-250MG
APOSOR927308-1GEA 632 EUR
APOSOR927308-1G APOSOR927308-250MG
Ethyl 3-(4-pyridyl)propiolate 95%
Ethyl 3-(4-pyridyl)propiolate
Formel: C₁₀H₉NO₂
Molecular Weight: 175,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23381103
CAS-Nummer: 66869-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR