2-Amino-4-chlorobenzaldehyde 95%

Lieferant: Apollo Scientific

OR927314-1G OR927314-250MG
APOSOR927314-1GEA 36.4 EUR
APOSOR927314-1G APOSOR927314-250MG
2-Amino-4-chlorobenzaldehyde 95%
2-Amino-4-chlorobenzaldehyde
Formel: C₇H₆ClNO
Molecular Weight: 155.582 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696881
CAS-Nummer: 59236-37-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR