2-Chloro-pyrido[2,3-b]pyrazine 95%
Lieferant: Apollo Scientific
Synonyme:
2-Chloropyrido[2,3-b]pyrazine
OR927315-1G
OR927315-250MG
APOSOR927315-1GEA
683
EUR
APOSOR927315-1G
APOSOR927315-250MG
2-Chloro-pyrido[2,3-b]pyrazine 95%
2-Chloro-pyrido[2,3-b]pyrazine
Formel:
C₇H₄ClN₃ Molecular Weight: 165,58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06738667 CAS-Nummer: 70838-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...