N-Ethyl-2-(1H-pyrazol-1-yl)ethanamine 95%

Lieferant: Apollo Scientific

OR927360-1G OR927360-5G
APOSOR927360-1GEA 319 EUR
APOSOR927360-1G APOSOR927360-5G
N-Ethyl-2-(1H-pyrazol-1-yl)ethanamine 95%
N-Ethyl-2-(1H-pyrazol-1-yl)ethanamine
Formel: C₇H₁₃N₃
Molecular Weight: 139.198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864293
CAS-Nummer: 340967-02-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR