1,2,3,4-Tetrahydroquinolin-5-amine
Lieferant: Apollo Scientific
|
Achtung
|
OR927400-1G
OR927400-250MG
APOSOR927400-1GEA
661
EUR
APOSOR927400-1G
APOSOR927400-250MG
1,2,3,4-Tetrahydroquinolin-5-amine
1,2,3,4-Tetrahydroquinolin-5-amine
Formel:
C₉H₁₂N₂ Molecular Weight: 148,205 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08234589 CAS-Nummer: 36887-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...