(5-Chlorothiophen-2-yl)methanol 95%

Lieferant: Apollo Scientific

OR927466-25G
APOSOR927466-25GEA 156 EUR
APOSOR927466-25G
(5-Chlorothiophen-2-yl)methanol 95%
(5-Chlorothiophen-2-yl)methanol
Formel: C₅H₅ClOS
Molecular Weight: 148,611 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06202649
CAS-Nummer: 74168-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR