3-Cyclopropyl-1-methyl-1H-pyrazole-4-carbaldehyde 95%
Lieferant: Apollo Scientific
OR927484-100MG
OR927484-1G
OR927484-250MG
APOSOR927484-100MGEA
183
EUR
APOSOR927484-100MG
APOSOR927484-1G
APOSOR927484-250MG
3-Cyclopropyl-1-methyl-1H-pyrazole-4-carbaldehyde 95%
3-Cyclopropyl-1-methyl-1H-pyrazole-4-carbaldehyde
Formel:
C₈H₁₀N₂O Molecular Weight: 150.178 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04970939 CAS-Nummer: 902837-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...