5-Amino-3-chloro-2-methoxypyridine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-6-methoxypyridin-3-amine
OR927679-25G
OR927679-5G
APOSOR927679-25GEA
652
EUR
APOSOR927679-25G
APOSOR927679-5G
5-Amino-3-chloro-2-methoxypyridine 98%
5-Amino-3-chloro-2-methoxypyridine
Formel:
C₆H₇ClN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD13190367 CAS-Nummer: 158387-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...