2-Aminocyclohex-1-ene-1-carbonitrile 96%
Lieferant: Apollo Scientific
Synonyme:
2-Aminocyclohex-1-enecarbonitrile
OR927699-1G
OR927699-250MG
OR927699-5G
APOSOR927699-1GEA
66.7
EUR
APOSOR927699-1G
APOSOR927699-250MG
APOSOR927699-5G
2-Aminocyclohex-1-ene-1-carbonitrile 96%
2-Aminocyclohex-1-ene-1-carbonitrile
Formel:
C₇H₁₀N₂ Molecular Weight: 122.168 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16046430 CAS-Nummer: 15595-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...