5-Aminomethyl-pyridine-2-carbonitrile 95%

Lieferant: Apollo Scientific

OR927724-100MG OR927724-1G OR927724-250MG
APOSOR927724-100MGEA 0 EUR
APOSOR927724-100MG APOSOR927724-1G APOSOR927724-250MG
5-Aminomethyl-pyridine-2-carbonitrile 95%
5-Aminomethyl-pyridine-2-carbonitrile
Formel: C₇H₇N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08457627
CAS-Nummer: 181130-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR