3-(Thien-3-yl)aniline 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Thiophen-3-yl)aniline, 3-(3-Aminophenyl)thiophene

OR9278-1G OR9278-250MG OR9278-5G
APOSOR9278-1GEA 265 EUR
APOSOR9278-1G APOSOR9278-250MG APOSOR9278-5G
3-(Thien-3-yl)aniline 97%
3-(Thien-3-yl)aniline
Formel: C₁₀H₉NS
Molecular Weight: 175,25 g/mol
Schmelzpunkt: 88,5…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06740169
CAS-Nummer: 161886-96-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR