Imidazo[1,2-a]pyridin-3-ylmethyl-methyl-amine 95%

Lieferant: Apollo Scientific

OR927830-1G OR927830-250MG
APOSOR927830-1GEA 683 EUR
APOSOR927830-1G APOSOR927830-250MG
Imidazo[1,2-a]pyridin-3-ylmethyl-methyl-amine 95%
Imidazo[1,2-a]pyridin-3-ylmethyl-methyl-amine
Formel: C₉H₁₁N₃
Molecular Weight: 161.204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739260
CAS-Nummer: 885275-83-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR