5-Chloroquinazoline-2,4(1H,3H)-dione 98%
Lieferant: Apollo Scientific
OR927915-1G
OR927915-25G
OR927915-5G
APOSOR927915-1GEA
42.2
EUR
APOSOR927915-1G
APOSOR927915-25G
APOSOR927915-5G
5-Chloroquinazoline-2,4(1H,3H)-dione 98%
5-Chloroquinazoline-2,4(1H,3H)-dione
Formel:
C₈H₅ClN₂O₂ Molecular Weight: 196,591 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09954845 CAS-Nummer: 78754-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...