1-Acetyl-3-hydroxyindole 98%

Lieferant: Apollo Scientific
Achtung

OR928060-25G OR928060-5G
APOSOR928060-25GEA 848 EUR
APOSOR928060-25G APOSOR928060-5G
1-Acetyl-3-hydroxyindole 98%
1-Acetyl-3-hydroxyindole
Formel: C₁₀H₉NO₂
Molecular Weight: 175,184 g/mol
Schmelzpunkt: 134…136 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00047261
CAS-Nummer: 33025-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR