1H-Indol-1-amine 95%
Lieferant: Apollo Scientific
Synonyme:
1-Aminoindole
OR928208-1G
OR928208-5G
APOSOR928208-1GEA
56.7
EUR
APOSOR928208-1G
APOSOR928208-5G
1H-Indol-1-amine 95%
1H-Indol-1-amine
Formel:
C₈H₈N₂ Molecular Weight: 132.163 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151983 CAS-Nummer: 53406-38-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...