Methyl 2-chloro-3-methylisonicotinate 97%
Lieferant: Apollo Scientific
OR928310-1G
OR928310-250MG
APOSOR928310-1GEA
78.3
EUR
APOSOR928310-1G
APOSOR928310-250MG
Methyl 2-chloro-3-methylisonicotinate 97%
Methyl 2-chloro-3-methylisonicotinate
Formel:
C₈H₈ClNO₂ Molecular Weight: 185,608 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08275065 CAS-Nummer: 787596-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...