6-Methoxy-7-hydroxyquinazolin-4-one 95%

Lieferant: Apollo Scientific
Achtung

OR928311-25G OR928311-5G
APOSOR928311-25GEA 238 EUR
APOSOR928311-25G APOSOR928311-5G
6-Methoxy-7-hydroxyquinazolin-4-one 95%
6-Methoxy-7-hydroxyquinazolin-4-one
Formel: C₉H₈N₂O₃
Molecular Weight: 192,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458441
CAS-Nummer: 162012-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR