Sulphachloropyrazine sodium 98%
Lieferant: Apollo Scientific
Synonyme:
Sodium ((4-aminophenyl)sulphonyl)(5-chloropyrazin-2-yl)amide
OR928389-1G
OR928389-5G
APOSOR928389-1GEA
284
EUR
APOSOR928389-1G
APOSOR928389-5G
Sulphachloropyrazine sodium 98%
Sulphachloropyrazine sodium
Formel:
C₁₀H₈ClN₄NaO₂S Molecular Weight: 306,704 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06798154 CAS-Nummer: 71720-40-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...