2-(3-Formylphenoxy)acetamide 95%

Lieferant: Apollo Scientific

OR928412-1G OR928412-5G
APOSOR928412-1GEA 132 EUR
APOSOR928412-1G APOSOR928412-5G
2-(3-Formylphenoxy)acetamide 95%
2-(3-Formylphenoxy)acetamide
Formel: C₉H₉NO₃
Molecular Weight: 179,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04374050
CAS-Nummer: 849015-95-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR