(2-Nitro-3-methoxy-phenyl)-methanol 95%

Lieferant: Apollo Scientific

Synonyme: (3-Methoxy-2-nitrophenyl)methanol

OR928421-1G OR928421-250MG
APOSOR928421-1GEA 140 EUR
APOSOR928421-1G APOSOR928421-250MG
(2-Nitro-3-methoxy-phenyl)-methanol 95%
(2-Nitro-3-methoxy-phenyl)-methanol
Formel: C₈H₉NO₄
Molecular Weight: 183,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06202768
CAS-Nummer: 53055-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR