4-Chloro-3H-[1,2,3]triazolo[4,5-c]pyridine 95%
Lieferant: Apollo Scientific
OR928520-1G
OR928520-250MG
APOSOR928520-1GEA
313
EUR
APOSOR928520-1G
APOSOR928520-250MG
4-Chloro-3H-[1,2,3]triazolo[4,5-c]pyridine 95%
4-Chloro-3H-[1,2,3]triazolo[4,5-c]pyridine
Formel:
C₅H₃ClN₄ Molecular Weight: 154,557 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13193520 CAS-Nummer: 36258-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...