6,7-Dihydro-5h-quinolin-8-one oxime 95%
Lieferant: Apollo Scientific
Synonyme: 6,7-Dihydroquinolin-8(5H)-one oxime
OR928605-1G OR928605-250MG
APOSOR928605-1GEA 339 EUR
APOSOR928605-1G APOSOR928605-250MG
6,7-Dihydro-5h-quinolin-8-one oxime 95%
6,7-Dihydro-5h-quinolin-8-one oxime
Formel: C₉H₁₀N₂O
Molecular Weight: 162,188 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20921531
CAS-Nummer: 58509-59-4