H-1,3-Cis-achc-ome hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme: cis-Methyl 3-aminocyclohexanecarboxylate hydrochloride
OR928883-1G OR928883-250MG
APOSOR928883-1GEA 154 EUR
APOSOR928883-1G APOSOR928883-250MG
H-1,3-Cis-achc-ome hydrochloride 95%
H-1,3-Cis-achc-ome hydrochloride
Formel: C₈H₁₅NO₂·ClH
Molecular Weight: 193,671 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13184930
CAS-Nummer: 87360-22-3