4-Nitro-5,6,7,8-tetrahydroquinolin-1-ium-1-olate 95%
Lieferant: Apollo Scientific
Synonyme:
4-Nitro-5,6,7,8-tetrahydroquinoline 1-oxide
OR928908-1G
OR928908-5G
APOSOR928908-1GEA
121
EUR
APOSOR928908-1G
APOSOR928908-5G
4-Nitro-5,6,7,8-tetrahydroquinolin-1-ium-1-olate 95%
4-Nitro-5,6,7,8-tetrahydroquinolin-1-ium-1-olate
Formel:
C₉H₁₀N₂O₃ Molecular Weight: 194,187 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20921528 CAS-Nummer: 125162-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...