5-(Pyridin-4-yl)-1,2-oxazole-3-carboxylic acid 95%

Lieferant: Apollo Scientific

OR928917-100MG OR928917-1G OR928917-250MG
APOSOR928917-100MGEA 161 EUR
APOSOR928917-100MG APOSOR928917-1G APOSOR928917-250MG
5-(Pyridin-4-yl)-1,2-oxazole-3-carboxylic acid 95%
5-(Pyridin-4-yl)-1,2-oxazole-3-carboxylic acid
Formel: C₉H₆N₂O₃
Molecular Weight: 190,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07377115
CAS-Nummer: 893638-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR