4-(2-Piperidyl)-1-butanol 95%

Lieferant: Apollo Scientific

OR928941-1G OR928941-250MG
APOSOR928941-1GEA 632 EUR
APOSOR928941-1G APOSOR928941-250MG
4-(2-Piperidyl)-1-butanol 95%
4-(2-Piperidyl)-1-butanol
Formel: C₉H₁₉NO
Molecular Weight: 157,253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13183976
CAS-Nummer: 90726-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR