Imidazo[1,2-a]pyrimidine-3-carbaldehyde 98%

Lieferant: Apollo Scientific

OR928950-1G OR928950-250MG
APOSOR928950-1GEA 91.5 EUR
APOSOR928950-1G APOSOR928950-250MG
Imidazo[1,2-a]pyrimidine-3-carbaldehyde 98%
Imidazo[1,2-a]pyrimidine-3-carbaldehyde
Formel: C₇H₅N₃O
Molecular Weight: 147,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027563
CAS-Nummer: 106012-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR