3-Allyl-4-ethoxybenzaldehyde 95%

Lieferant: Apollo Scientific

OR928959-1G OR928959-5G
APOSOR928959-1GEA 219 EUR
APOSOR928959-1G APOSOR928959-5G
3-Allyl-4-ethoxybenzaldehyde 95%
3-Allyl-4-ethoxybenzaldehyde
Formel: C₁₂H₁₄O₂
Molecular Weight: 190,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08691768
CAS-Nummer: 915922-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR