1-Cyclopentyl-1H-pyrrole-3-carbaldehyde 95%
Lieferant: Apollo Scientific
OR928965-1G
OR928965-5G
APOSOR928965-1GEA
219
EUR
APOSOR928965-1G
APOSOR928965-5G
1-Cyclopentyl-1H-pyrrole-3-carbaldehyde 95%
1-Cyclopentyl-1H-pyrrole-3-carbaldehyde
Formel:
C₁₀H₁₃NO Molecular Weight: 163.216 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11695056 CAS-Nummer: 1071359-81-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...