4-Bromo-2-ethynylpyridine 98%
Lieferant: Apollo Scientific
OR928998-1G
OR928998-250MG
APOSOR928998-1GEA
632
EUR
APOSOR928998-1G
APOSOR928998-250MG
4-Bromo-2-ethynylpyridine 98%
4-Bromo-2-ethynylpyridine
Formel:
C₇H₄BrN Molecular Weight: 182.017 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13189721 CAS-Nummer: 1196155-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...