4-Bromo-1,3-dihydro-2-benzofuran 95%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-1,3-dihydroisobenzofuran

OR929157-1G OR929157-250MG
APOSOR929157-1GEA 114 EUR
APOSOR929157-1G APOSOR929157-250MG
4-Bromo-1,3-dihydro-2-benzofuran 95%
4-Bromo-1,3-dihydro-2-benzofuran
Formel: C₈H₇BrO
Molecular Weight: 199,045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22397733
CAS-Nummer: 1402667-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR