5-Bromo-L-tryptophan 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-2-Amino-3-(5-bromo-1H-indol-3-yl)propanoic acid, H-Trp(5-Br)-OH
OR929180-1G
OR929180-250MG
OR929180-500MG
APOSOR929180-1GEA
112
EUR
APOSOR929180-1G
APOSOR929180-250MG
APOSOR929180-500MG
5-Bromo-L-tryptophan 97%
5-Bromo-L-tryptophan
Formel:
C₁₁H₁₁BrN₂O₂ Molecular Weight: 283,121 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06796707 CAS-Nummer: 25197-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...