2-Chloro-5-methoxypyridin-3-amine 96%

Lieferant: Apollo Scientific
Achtung

OR929197-1G OR929197-250MG
APOSOR929197-1GEA 115 EUR
APOSOR929197-1G APOSOR929197-250MG
2-Chloro-5-methoxypyridin-3-amine 96%
2-Chloro-5-methoxypyridin-3-amine
Formel: C₆H₇ClN₂O
Molecular Weight: 158.586 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11044248
CAS-Nummer: 720666-45-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR