Acetyl-d-alloisoleucine 95%

Lieferant: Apollo Scientific

Synonyme: N-Acetyl-D-allo-isoleucine

OR929224-1G OR929224-250MG
APOSOR929224-1GEA 121 EUR
APOSOR929224-1G APOSOR929224-250MG
Acetyl-d-alloisoleucine 95%
Acetyl-d-alloisoleucine
Formel: C₈H₁₅NO₃
Molecular Weight: 173,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00136538
CAS-Nummer: 54831-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR