2-Chloro-3-methylisonicotinic acid 98%
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-3-methyl-4-pyridinecarboxylic Acid
OR929238-1G
OR929238-250MG
APOSOR929238-1GEA
310
EUR
APOSOR929238-1G
APOSOR929238-250MG
2-Chloro-3-methylisonicotinic acid 98%
2-Chloro-3-methylisonicotinic acid
Formel:
C₇H₆ClNO₂ Molecular Weight: 171,581 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16610679 CAS-Nummer: 133928-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...