(R)-formamido(phenyl)acetic acid 95%

Lieferant: Apollo Scientific

OR929359-1G OR929359-250MG
APOSOR929359-1GEA 502 EUR
APOSOR929359-1G APOSOR929359-250MG
(R)-formamido(phenyl)acetic acid 95%
(R)-formamido(phenyl)acetic acid
Formel: C₉H₉NO₃
Molecular Weight: 179,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03093479
CAS-Nummer: 10419-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR