1-(2-Methylphenyl)-1H-pyrazole-4-carbaldehyde 98%
Lieferant: Apollo Scientific
OR929449-10G
OR929449-1G
OR929449-5G
APOSOR929449-10GEA
647
EUR
APOSOR929449-10G
APOSOR929449-1G
APOSOR929449-5G
1-(2-Methylphenyl)-1H-pyrazole-4-carbaldehyde 98%
1-(2-Methylphenyl)-1H-pyrazole-4-carbaldehyde
Formel:
C₁₁H₁₀N₂O Molecular Weight: 186,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03780730 CAS-Nummer: 75815-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...