4,6-Dichloro-1-methyl-1H-imidazo[4,5-c]pyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR929468-100MG
OR929468-1G
OR929468-250MG
APOSOR929468-100MGEA
39.3
EUR
APOSOR929468-100MG
APOSOR929468-1G
APOSOR929468-250MG
4,6-Dichloro-1-methyl-1H-imidazo[4,5-c]pyridine 95%
4,6-Dichloro-1-methyl-1H-imidazo[4,5-c]pyridine
Formel:
C₇H₅Cl₂N₃ Molecular Weight: 202.041 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21603853 CAS-Nummer: 887147-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...