2-(Bromoethyl)methylsulphone 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-2-(methylsulphonyl)ethane, 2-Bromoethyl Methyl sulphone

OR929569-1G OR929569-5G
APOSOR929569-1GEA 34.8 EUR
APOSOR929569-1G APOSOR929569-5G
2-(Bromoethyl)methylsulphone 98%
2-(Bromoethyl)methylsulphone
Formel: C₃H₇BrO₂S
Molecular Weight: 187,055 g/mol
Schmelzpunkt: 32…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08274926
CAS-Nummer: 16523-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR