5-Chloro-n,n-dimethylpyridin-2-amine 95%

Lieferant: Apollo Scientific

OR929595-1G OR929595-250MG
APOSOR929595-1GEA 567 EUR
APOSOR929595-1G APOSOR929595-250MG
5-Chloro-n,n-dimethylpyridin-2-amine 95%
5-Chloro-n,n-dimethylpyridin-2-amine
Formel: C₇H₉ClN₂
Molecular Weight: 156,613 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13188972
CAS-Nummer: 26163-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR