Fmoc-O-sulpho-L-tyrosine sodium salt 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S)-2-[[9H-fluoren-9-ylmethoxy(hydroxy)methyl]amino]-3-(4-sulphooxyphenyl)propanoate sodium salt

OR929796-1G OR929796-250MG
APOSOR929796-1GEA 982 EUR
APOSOR929796-1G APOSOR929796-250MG
Fmoc-O-sulpho-L-tyrosine sodium salt 97%
Fmoc-O-sulpho-L-tyrosine sodium salt
Formel: C₂₄H₂₂NO₈S·Na
Molecular Weight: 507,488 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00077078
CAS-Nummer: 106864-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR