1-(2-Methoxyethyl)-1H-pyrrole-2-carbaldehyde 95%
Lieferant: Apollo Scientific
OR930046-1G
OR930046-250MG
APOSOR930046-1GEA
284
EUR
APOSOR930046-1G
APOSOR930046-250MG
1-(2-Methoxyethyl)-1H-pyrrole-2-carbaldehyde 95%
1-(2-Methoxyethyl)-1H-pyrrole-2-carbaldehyde
Formel:
C₈H₁₁NO₂ Molecular Weight: 153,178 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09674980 CAS-Nummer: 573720-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...