Methyl 5-acetylthiophene-2-carboxylate 98%
Lieferant: Apollo Scientific
OR930150-1G
OR930150-5G
APOSOR930150-1GEA
142
EUR
APOSOR930150-1G
APOSOR930150-5G
Methyl 5-acetylthiophene-2-carboxylate 98%
Methyl 5-acetylthiophene-2-carboxylate
Formel:
C₈H₈O₃S Molecular Weight: 184,212 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06204049 CAS-Nummer: 4101-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...